Skip to Content
Merck

PG9H9BC7B16D

PrecisePEG

Fmoc-Gly-Gly-L-Phe-N-[(carboxymethoxy)methyl]Glycinamide

Synonym(s):

(S)-11-benzyl-1-(9H-fluoren-9-yl)-3,6,9,12,15-pentaoxo-2,18-dioxa-4,7,10,13,16-pentaazaicosan-20-oic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C33H35O9N5
CAS Number:
Molecular Weight:
645.67
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C33H35N5O9/c39-28(16-36-33(45)47-18-26-24-12-6-4-10-22(24)23-11-5-7-13-25(23)26)34-17-30(41)38-27(14-21-8-2-1-3-9-21)32(44)35-15-29(40)37-20-46-19-31(42)43/h1-13,26-27H,14-20H2,(H,34,39)(H,35,44)(H,36,45)(H,37,40)(H,38,41)(H,42,43)/t27-/m0/s1

Inchi Key

JOZGCSZISBFCOO-MHZLTWQESA-N

Shipping Temperature

ambient

Purity

96%

Storage temp.

-20

Country of Origin

CN

Physical Form

Solid



hstatements

Precautionary Statements



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library