Skip to Content
Merck

RIE156324326

Rieke Metals, Inc.

3,4-difluoro-4'-piperidinomethyl benzophenone

Synonym(s):

(3,4-difluorophenyl)[4-(1-piperidinylmethyl)phenyl]methanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
315.36
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C19H19F2NO/c20-17-9-8-16(12-18(17)21)19(23)15-6-4-14(5-7-15)13-22-10-2-1-3-11-22/h4-9,12H,1-3,10-11,13H2

Inchi Key

KMPUPAJNUYLMFT-UHFFFAOYSA-N

Purity

97%

Country of Origin

US



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library