Skip to Content
Merck

Skip To

IRIS-1

Isotopic certified reference material of natural iridium

certified reference material

Synonym(s):

Isotopic certified reference material of natural iridium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
eCl@ss:
25140904
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

agency

traceable to (National Research Council of Canada (NRC))

manufacturer/tradename

National Research Council of Canada (NRC)

application(s)

food and beverages

format

neat

storage temp.

room temp

General description

NRC Canada’s certified reference materials (CRMs) are traceable to the International System of Units, and conform to the requirements of ISO/IEC 17025 and ISO 17034.For specific values, as well as additional information regarding stability and storage information, please view the Certificate of Analysis.With NRC Canada’s CRMs, you gain access to products backed by extensive research and expertise, fostering trust and consistency in your results. Elevate your laboratory’s accuracy and credibility with NRC Canada’s certified reference materials.
Certified values for isotope amount ratios, isotope abundances and atomic weight of iridium have been established for this isotopic reference material (IRIS-1). For more information, please refer to the Certificate of Analysis or Safety Data Sheet.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
LUIS1OSIS1HALF1
description

certified reference material

description

certified reference material

description

certified reference material

description

certified reference material

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages

format

neat

format

neat

format

neat

format

single component solution

agency

traceable to (National Research Council of Canada (NRC))

agency

traceable to (National Research Council of Canada (NRC))

agency

-

agency

-

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

2-8°C

manufacturer/tradename

National Research Council of Canada (NRC)

manufacturer/tradename

National Research Council of Canada (NRC)

manufacturer/tradename

National Research Council of Canada (NRC)

manufacturer/tradename

National Research Council of Canada (NRC)


Pictograms

Corrosion

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library