Skip to Content
Merck

Skip To

PHR8564

Nitroso aryl piperazine solution

1 mg/mL in methanol, ampule of 1 mL, certified reference material

Synonym(s):

11-(4-nitrosopiperazin-1-yl)dibenzo[b,f][1,4]thiazepine, Quetiapine EP Impurity B Nitroso Analog, N-Nitroso Aryl Piperazine Quatiapine, Quetiapine nitroso intermediate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H16N4OS
Molecular Weight:
324.40
UNSPSC Code:
41116107
NACRES:
NA.21
eCl@ss:
25140904
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

agency

(Not traceable to any primary pharmacopeia standard)

API family

quetiapine

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1 mg/mL in methanol

application(s)

pharmaceutical

format

single component solution

storage temp.

-10 to -25°C

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1592715
description

1 mg/mL in methanol, ampule of 1 mL, certified reference material

description

United States Pharmacopeia (USP) Reference Standard

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)
pharmaceutical
pharmaceutical

format

single component solution

format

neat

grade

certified reference material

grade

pharmaceutical primary standard

agency

(Not traceable to any primary pharmacopeia standard)

agency

USP

storage temp.

-10 to -25°C

storage temp.

2-8°C

concentration

1 mg/mL in methanol

concentration

-


Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

Flash Point (°F)

49.5 °F - closed cup

Flash Point (°C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-56-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library