Skip to Content
Merck

APREST71270

PrEST Antigen ARFGEF2

Prestige Antigens™ Powered by Atlas Antibodies, buffered aqueous solution

Synonym(s):

BIG2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli

assay

>80% (SDS-PAGE)

form

buffered aqueous solution

mol wt

predicted mol wt 31 kDa

purified by

immobilized metal affinity chromatography (IMAC)

concentration

≥0.5 mg/mL

immunogen sequence

KPIQSKPQSPVIQAAAVSPKFVRLKHSQAQSKPTTPEKTDLTNGEHARSDSGKVSTENGDAPRERGSSLSGTDDGAQEVVKDILEDVVTSAIKAAEKHGLTEPERVLGELECQECAIPPGVDENSQTNGI

Ensembl | human accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ARFGEF2(10564)

General description

Recombinant protein fragment of Human ARFGEF2 with N-terminal His6ABP (ABP = Albumin Binding Protein derived from Streptococcal Protein G) fusion tag.

Application

Suitable as a blocking agent using corresponding antibodies.

Physical form

Solution in 1 M urea-PBS, pH 7.4

Preparation Note

The protein solution should be gently mixed before use. Optimal concentrations and conditions for each application should be determined by the user.

Other Notes

Corresponding Antibody HPA026078.

Legal Information

Prestige Antigens is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

10 - Combustible liquids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library