Skip to Content
Merck

BPS72048

ARG1 Inhibitor Screening Assay Kit

Synonym(s):

Assay Kits, Arginase 1, LCN7

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41121811
eCl@ss:
32160701
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

BPS Bioscience

technique(s)

activity assay: suitable (Colorimetric - UV Spectrophotometry)

UniProt accession no.

shipped in

dry ice

storage temp.

-65 to -96°C

General description

The ARG1 Inhibitor Screening Assay Kit is designed to measure ARG1 enzyme inhibition. The kit comes in a convenient format, with enough reaction solution and enzyme to perform a total of 100 reactions. The ARG1 Inhibitor Screening Assay Kit is simple to use. Inhibitor and enzyme are added to a sample containing thioarginine substrate. After a room temperature incubation, activity is determined by measuring the absorption of reaction product at λ=410-415 nm.

Application

Study enzyme kinetics and screen small molecule inhibitors for drug discovery in high throughput screening (HTS) applications


Arginase enzymes convert arginine to ornithine through hydrolysis. Two known isoforms of Arginase exist, ARG1 and ARG2. This enzyme is involved in the regulation of a variety of immunological responses and is a major target in immunotherapy. ARG1/2 is overexpressed in myeloid-derived suppressor cells (MDSCs) and tumor-associated macrophages (TAMs). Overexpression of ARG1/2 results in depleted levels of arginine both intracellularly and extracellularly. As arginine levels are depleted in the microenvironment, immune cells are starved of this amino acid and the function of key immunological activators become impaired; T cell proliferation is inhibited, regulatory T cells become activated and inhibit CD4+ T cells, and immunosuppressants have increased longevity. Depleted arginine also results in the release of reactive nitrogen species and reactive oxygen species from TAMs and MDSCs. These reactive species cause T cell apoptosis and the activation and growth of antigen presenting cells.

Datasheet and Protocols for the 96 reaction kit may be found here.

Datasheet and Protocols for the 384 reaction kit may be found here.


Signal Word

Warning

Pictograms

Exclamation mark

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library