Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
47101613
Pricing and availability is not currently available.
manufacturer/tradename
BPS Bioscience
storage temp.
-65 to -96°C
General description
This buffer is intended for use with mammalian cells during magnetic cell isolation or single cell processing. This buffer is optimized and formulated to reduce nonspecific binding and cell clumping
Application
Preparation Note
Prepared under sterile conditions and filtered (0.2 µm).Dilute 10-fold with sterile distilled water. For sterile conditions, filter the buffer after dilution and use under laminar flow hood.
Analysis Note
Avoid freeze/thaw cycles.
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description BioUltra, Molecular Biology, pH 7.8 | description 1.0 M, pH 7.4 (25 °C) |
| storage temp. -65 to -96°C | storage temp. - | storage temp. - | storage temp. 2-8°C |
| manufacturer/tradename BPS Bioscience | manufacturer/tradename Chemicon® | manufacturer/tradename - | manufacturer/tradename - |
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



