Skip to Content
Merck

Skip To

BPS82130

ADP-Ribosylation Cycle Inhibitor Mix

Synonym(s):

cycle inhibitor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

BPS Bioscience

storage temp.

-65 to -96°C

General description

The ADP-Ribosylation Cycle Inhibitor Mix is a 500x cocktail of two inhibitors (AZD2281 and PDD00017273). AZD2281 (Olaparib) is a PARP inhibitor with broad pan-PARP activity, which blocks a large proportion of the ADP-ribosylation of proteins. PDD00017273 is a potent, selective poly (ADP-ribose) glycohydrolase (PARG) inhibitor that blocks PARG-mediated de-ribosylation of proteins. Used together, these two inhibitors "freeze" the state of cellular ribosylation at the time the mix is applied to the cells. This product is designed to supplement a cell lysis buffer of choice to prevent post-lysis changes in the ADP-ribosylation status of extracted proteins.

Application

Datasheet and Protocols may be found here.

Preparation Note

Thaw right before use. Dilute 500-fold directly in the cell lysis buffer of choice. Aliquots are single use and should be discarded after thawing. Discard excess lysis buffer containing the inhibitor mix at the end of the day.Note: This inhibitor mix is designed for the analysis of proteins that are ribosylated by PARP1/2/3/4 and PARP10 (approximately 80% of proteins) and are de-ribosylated by PARG. For proteins that are the target of other PARPs or a ribosylation eraser other than PARG, different inhibitors may be necessary.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MABC547MABE1031A9106
description

-

description

clone 10H, from mouse

description

Anti-poly-ADP-ribose binding reagent is a reagent that selectively binds to ADP ribose for use in Western Blotting, Immunocytochemistry and Dot Blot.

description

1000-3000 units/mg protein

storage temp.

-65 to -96°C

storage temp.

-

storage temp.

-

storage temp.

−20°C

manufacturer/tradename

BPS Bioscience

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-


Storage Class

10 - Combustible liquids

Flash Point (°F)

188.6 °F - closed cup

Flash Point (°C)

87 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library