Skip to Content
Merck

Skip To

D1007

Dehydroisoandrosterone 3-glucuronide

Synonym(s):

5-Androsten-3β-ol-17-one 3-glucuronide, De­hydro­epi­andro­sterone 3-glucuronide, Pra­sterone glucur­onide

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C25H36O8
CAS Number:
Molecular Weight:
464.55
UNSPSC Code:
12352202
PubChem Substance ID:
MDL number:
eCl@ss:
32150313
Form:
solid


form

solid

drug control

Home Office Schedule 4.2; regulated under CDSA - not available from Sigma-Aldrich Canada

shipped in

ambient

storage temp.

−20°C

SMILES string

CC12CCC3C(CC=C4CC(CCC34C)OC5OC(C(O)C(O)C5O)C(O)=O)C1CCC2=O

InChI

1S/C25H36O8/c1-24-9-7-13(32-23-20(29)18(27)19(28)21(33-23)22(30)31)11-12(24)3-4-14-15-5-6-17(26)25(15,2)10-8-16(14)24/h3,13-16,18-21,23,27-29H,4-11H2,1-2H3,(H,30,31)

InChI key

GLONBVCUAVPJFV-UHFFFAOYSA-N



Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters