Skip to Content
Merck

Skip To

DNB300

GenElute™ Urine Cell-Free DNA Purification Mini Kit

Sustainability features

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.52
UNSPSC Code:
41105600


sustainability

Sustainability features

shipped in

ambient

storage temp.

room temp

General description

Sigma′s GenElute™ Urine Cell-Free DNA Purification Mini Kit provides a fast, reliable and simple procedures for isolating cell-free circulating DNA (cfc-DNA) from various amounts of urine ranging from 250 μL up to 2 mL. Purification is based on spin column chromatography that uses Sigma′s proprietary resin separation matrix. The kits are designed to isolate all sizes of cfc-DNA from either fresh or frozen urine samples. Moreover, these kits allow the user to elute the purified cfc-DNA into a flexible elution volume ranging from 25 μL to 50 μL. The purified urine cfc-DNA is eluted in an elution buffer that is compatible with all downstream applications including PCR, qPCR, methylation-sensitive PCR and Southern Blot analysis, and NGS.

  • Isolate all sizes of circulating DNA from fresh, preserved or frozen urine samples
  • Isolate viral DNA
  • Versatile urine input volumes (250 μL - 2 mL)
  • Concentrate circulating DNA into a flexible elution volume ranging from (50 μL - 100 μL)
  • Isolate inhibitor-free cell-free circulating DNA
  • Purify high-quality DNA in 15-20 minutes

Application

  • PCR
  • qPCR
  • Southern blot
  • Methylation-sensitive PCR
  • CpG array
  • Restriction enzyme digestions
  • Viral detection
  • Bacterial detection
  • Microarray
  • NGS

Other Notes

This product aligns with one or more of the 12 Principles of Green ChemistryThis product has Inherently Safer Chemistry, compared to the standard use of phenol and chloroform to perform DNA extractions.
Preps

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC


Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Met. Corr. 1 - Resp. Sens. 1 - Skin Irrit. 2

WGK

WGK 3

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters