Skip to Content
Merck

Skip To

F0807

Anti-FNTB (276-290) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-FPTB, Anti-Farnesyltransferase subunit β

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203


biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~49 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... FNTB(2342)

Immunogen

synthetic peptide corresponding to amino acids 276-290 of human FNTB

Application

Anti-FNTB (276-290) antibody produced in rabbit is suitable for western blotting at a dilution of 1:500-1:2,000.
Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Biochem/physiol Actions

Protein farnesyltransferase subunit β is an enzyme encoded by the FNTB gene in humans and is located on human chromosome bands 14q23-q24. It helps in catalyzing the biologically relevant lipidation of cellular proteins. FTase may play an important role in the genesis and development of primary liver cancer (PLC). It may act as a reliable marker for the metastatic activity gained by liver tumor cells. FTase may be used clinically in predicting metastatic recurrence of PLC. Farnesyl protein transferase (FPTase) inhibitors (FTIs) were also developed as potential anticancer agents targeting the ras oncogene and also for parasitic infections.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.


Still not finding the right product?

Try our Product Selector Tool to narrow your options




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters