Skip to Content
System maintenance is scheduled for 11 Sep, 08:00 PM - 12 Sep, 00:00 AM CST [12 Sep 03:00 AM to 07:00 AM CET] and will affect some web and offline transactions, including quotes, price checks, and order status inquiries. We apologize for any inconvenience.
Merck
Skip To
GE11-0012-54
Tricorn 10 Coarse Filter Kit
Sign In to View Organizational & Contract Pricing.
About This Item
UNSPSC Code:
41106500
description
Tricorn glass columns may be used with aqueous buffer solutions and detergents at pH conditions between 1 and 14, and with nearly all organic solutions commonly used in chromatography
feature
Kit Contents: 5 x Top filter 5 x Bottom filter 5 x O-ring 7 x 1 mm (EPDM) , Wetted Materials: Ethylene propylene diene (EPDM), Polyethylene (PE)
manufacturer/tradename
Cytiva 11-0012-54
parameter
max. temp. (60 °C), 4 °C min. temp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)