Skip to Content
Merck

Skip To

MTCD309643

MTC™ Bio OptiWell™ Deep Well Plate

96 square wells, U-bottom wells, H-style, capacity 2.2 mL, non-sterile, case of 50 ea (10 bags of 5ea)

Synonym(s):

automated research plate, MTC Bio D3096-43, automated assay plate, lab automation plate, research automation tray, robotic research plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41121800
eCl@ss:
32160702
Material:
U-bottom wells, clear plate, clear wells, polypropylene , square wells
Size:
96 wells
Sterility:
non-sterile
Feature:
lid: no, skirt: no
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

U-bottom wells, clear plate, clear wells, polypropylene , square wells

sterility

non-sterile

feature

lid: no, skirt: no

packaging

case of 50 ea (10 bags of 5ea)

manufacturer/tradename

MTC™ Bio D3096-43

parameter

4,000 × g speed

capacity

2.2 mL

size

96 wells

well volume

2.2 mL

General description

OptiWell plates come in 24, 48, 96 or 384 format for virtually every research lab, HTS application and automated/robotic platform. These plates are useful for bacterial cultures, centrifugation (up to 4000 x g) and general sample storage. Plates are produced/packaged in a Class 100,000 clean room and are ANSI / SLAS 1-2004 compliant. Several plates are sterilized by gamma irradiation and all are certified non-pyrogenic, RNase/DNase free.

Features and Benefits

  • For automated research platforms / robotic equipment
  • Produced/packaged in a Class 100,000 clean room
  • ANSI / SLAS 1-2004 compliant
  • Minimal binding / liquid retention
  • Black printed alpha-numerics
  • Virgin, medical-grade polypropylene
  • Sterilized by gamma irradiation
  • Certified non-pyrogenic, RNase/DNase free

Legal Information

MTC is a trademark of MTC Bio
OptiWell is a trademark of MTC Bio

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MTCD309645MTCD309635MTCD309637
description

96 square wells, U-bottom wells, H-style, capacity 2.2 mL, non-sterile, case of 50 ea (10 bags of 5ea)

description

96 square wells, V-bottom wells, capacity 2.3 mL, non-sterile, case of 50 ea (10 bags of 5ea)

description

96 round wells, U-bottom wells, capacity 2.0 mL, non-sterile, case of 50 ea (10 bags of 5ea)

description

96 round wells, U-bottom wells, capacity 2.0 mL, non-sterile, case of 50 ea (10 bags of 5ea)

manufacturer/tradename

MTC™ Bio D3096-43

manufacturer/tradename

MTC™ Bio D3096-45

manufacturer/tradename

MTC™ Bio D3096-35

manufacturer/tradename

MTC™ Bio D3096-37

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

feature

lid: no, skirt: no

feature

skirt: no, lid: no

feature

lid: no, skirt: no

feature

lid: no, skirt: no

material

U-bottom wells, clear plate, polypropylene , clear wells, square wells

material

clear wells, V-bottom wells, square wells, clear plate, polypropylene

material

U-bottom wells, clear wells, round wells, clear plate, polypropylene

material

polypropylene , clear wells, U-bottom wells, round wells, clear plate

size

96 wells

size

96 wells

size

96 wells

size

96 wells

packaging

case of 50 ea (10 bags of 5ea)

packaging

case of 50 ea (10 bags of 5ea)

packaging

case of 50 ea (10 bags of 5ea)

packaging

case of 50 ea (10 bags of 5ea)


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library