Skip to Content
Merck

Skip To

OGS165

PSF-T7-NH2-OMPA SP-NCOI - BACTERIAL SECRETION PLASMID

plasmid vector for molecular cloning

Synonym(s):

cloning vector, expression vector, molecular cloning vector, plasmid, plasmid vector, snapfast vector, vector

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.85
UNSPSC Code:
12352200
Promoter:
Promoter name: T7
Promoter activity: inducible
Promoter type: phage
Origin of replication:
pUC (500 copies)
Bacteria selection:
kanamycin
Reporter gene:
none
Peptide cleavage:
no cleavage


form

buffered aqueous solution

mol wt

size 3900 bp

bacteria selection

kanamycin

origin of replication

pUC (500 copies)

peptide cleavage

no cleavage

peptide tag location

N-terminal

promoter

Promoter name: T7
Promoter activity: inducible
Promoter type: phage

reporter gene

none

secretion signal

OmpA

shipped in

ambient

storage temp.

−20°C

General description

This vector allows transcription of un-polyadenylated and un-capped RNA using the T7 bacteriophage polymerase. This vector also contains the Escherichia coli (E. coli) outer membrane protein A (OmpA) secretory signal peptide. When positioned at the N-terminus of a protein it should direct the protein to be secreted out of the bacterial cytosol and into the periplasmic space. The OmpA secretory tag is 21 amino acids in length. The signal peptide protein sequence is MKKTAIAIAVALAGFATVAQA. Cleavage occurs after the final Alanine residue. The peptide tag will be cleaved the mature protein during export. In this vector the signal peptide coding sequence has been positioned adjacent to the NcoI restriction site.

This technique can be used to increase di-sulphide bond formation and also reduces proteolytic degradation. This method also reduces contamination from endogenous proteins because very few E. coli proteins are secreted. A sub fraction of the protein secreted into the periplasmic space can also extracted from the growth media. It is still unclear why this occurs but it believed to reflect increased permeability of the outer membrane particularly during extended culture periods.

Secretion of proteins in E. coli can sometimes be difficult. Potential issues include protein toxicity within the periplasm variable secretion efficiency formation of inclusion bodies within the cytoplasm when using strong promoters incorrect dis-sulphide formation. Background to bacterial secretion systems including possible solutions to these problems can be found in Choi et al 2004 Applied Microbiology and Biotechnology 64 (625-635).

all our vectors (except those in the terminators category on our website) will contain a T7 terminator downstream of the multiple cloning site. This will enable transcription termination.

Promoter Expression Level:

Application

Cloning in a gene: This vector has been designed to allow the addition of a peptide tag to the end of a protein of interest using standard cloning techniques.Multiple Cloning Site Notes:

There is a start codon in the NcoI site can be removed by digestion with KpnI if required. The MCS for gene insertions extends from NotI to XbaI however the tag resides between the NotI and HindIII sites. There are Shine-Dalgarno sequences and KOZAK sequences aligned with the start codon of the peptide tag.

The ClaI to NheI sites have other functions such as adding C-terminal peptide tags second promoters or IRES expression components. The BsgI and BseRI restriction sites cleave within the stop codon in the XbaI site and allow the retrospective fusion of C-terminal peptide tags sequences if the stop codon is placed in this position.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.oxgene.com

Other Notes

To view sequence information for this product, please visit the product page


Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters