Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.85
UNSPSC Code:
12352200
eCl@ss:
32160703
Promoter:
Promoter name: OXB20
Promoter activity: constitutive
Promoter type: bacterial
Promoter activity: constitutive
Promoter type: bacterial
Origin of replication:
pUC (500 copies)
Bacteria selection:
kanamycin
Reporter gene:
none
Peptide cleavage:
no cleavage
Pricing and availability is not currently available.
recombinant
expressed in E. coli
tag
10-His tagged
form
buffered aqueous solution
mol wt
size 3872 bp
technique(s)
molecular cloning: suitable
bacteria selection
kanamycin
origin of replication
pUC (500 copies)
peptide cleavage
no cleavage
peptide tag location
C-terminal
promoter
Promoter name: OXB20
Promoter activity: constitutive
Promoter type: bacterial
reporter gene
none
shipped in
ambient
storage temp.
−20°C
General description
This plasmid is designed to express tagged proteins in E. coli. The plasmid contains a constitutive promoter (OXB20) derived from the region upstream of the E. coli RecA gene. It does not require induction or any additional components for activity. It is the strongest of the bacterial promoters that we provide and this high level of expression can cause expression problems with some proteins with poor solubility. For this reason we sell a range of bacterial promoters with different expression levels (OXB1(low)>OXB20(high)) that can be provided with the peptide tags in this plasmid on request.
About the Peptide Tag:This plasmid contains a c-terminal Hexa-Histidine (6His) reporter tag that can be fused to a gene of interest to allow protein detection and/or purification. The sequence of the tag is: HHHHHH.
About the Cleavage Tag:This plasmid does not contain a protease cleavage site.
Promoter Expression Level: This plasmid contains a constitutive bacterial promoter that does not require induction. It is the strongest bacterial promoter we sell and this can cause solubility and expression problems with some proteins. We also offer a range of other bacterial promoters that are compatible with this plasmid and are available on request.
About the Peptide Tag:This plasmid contains a c-terminal Hexa-Histidine (6His) reporter tag that can be fused to a gene of interest to allow protein detection and/or purification. The sequence of the tag is: HHHHHH.
About the Cleavage Tag:This plasmid does not contain a protease cleavage site.
Promoter Expression Level: This plasmid contains a constitutive bacterial promoter that does not require induction. It is the strongest bacterial promoter we sell and this can cause solubility and expression problems with some proteins. We also offer a range of other bacterial promoters that are compatible with this plasmid and are available on request.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.oxfordgenetics.com.
Other Notes
Looking for more vector options to move your experiments forward faster? Consider a custom cloning vector designed and built by Oxford Genetics™. Find out more at Oxford Genetics - Sigma's partner for cloning and expression vectors for molecular biology and synthetic biology applications.
Please select the file type you require. For reference most cloning programs will import a .gb (Genbank) file and will show all of the plasmids features automatically when downloaded and imported.
Genebank Vector Sequence File
FASTA Vector Sequence File
Full Plasmid Map
Genebank Vector Sequence File
FASTA Vector Sequence File
Full Plasmid Map
Legal Information
Oxford Genetics is a trademark of Oxford Genetics Ltd
Storage Class
12 - Non Combustible Liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.