Skip to Content
Merck

SML0168

Fluorescent Histamine H3/H4 receptor Antagonist (H3-633-AN) synthetic

Sign In to View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C44H48BF2N8O3S2
Molecular Weight:
849.84
UNSPSC Code:
12352200
NACRES:
NA.77

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

storage temp.

−20°C

Quality Level

Biochem/physiol Actions

The SML0168 ligand was shown to antagonize the activity of the histamine agonist, histamine, in two separate recombinant CHO cell lines expressing either the human H2 or H3-receptor and a cyclic-AMP responsive secreted placental alkaline phosphatase (SPAP) reporter gene.

Features and Benefits

This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

Dissolve 0.2 mg in 23.5 ul of DMSO to give a 10mM stock solution. Once reconstituted into DMSO the product, must be stored, preferably in smaller aliquots, at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offers many products related to histamine receptors for your research needs.

Related Content

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service