Sign In to View Organizational & Contract Pricing
Select a Size
About This Item
Empirical Formula (Hill Notation):
C16H17F2N5O2
CAS Number:
Molecular Weight:
349.34
NACRES:
NA.28
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
2-8°C
SMILES string
O=C(NC1CCNCC1)C2=NNC=C2NC(C3=CC(F)=CC(F)=C3)=O
Biochem/physiol Actions
Potent cyclin-dependent kinase-like 5 (CDKL5) and glycogen synthase kinase-3 (GSK3⍺/β) inhibitor with good kinome-wide selectivity.
SGC-CDKL5/GSK3-1 is a potent cyclin-dependent kinase-like 5 and glycogen synthase kinase-3 inhibitor (CDKL5/GSK3⍺/GSK3β IC50 = 6.5/4/9 nM by cell-free enzymatic assay, 3.5/10/35 nM by cellular NanoBRET) with good kinome-wide selectivity. SGC-CDKL5/GSK3-1 protects iPSC-derived motor neurons from ER stress-induced death.
SGC-CDKL5/GSK3-1 is a potent cyclin-dependent kinase-like 5 and glycogen synthase kinase-3 inhibitor (CDKL5/GSK3⍺/GSK3β IC50 = 6.5/4/9 nM by cell-free enzymatic assay, 3.5/10/35 nM by cellular NanoBRET) with good kinome-wide selectivity. SGC-CDKL5/GSK3-1 protects iPSC-derived motor neurons from ER stress-induced death.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service