Skip to Content
Merck

UPBST-RTB

Ultra pure BST-RT Reaction Buffer

Suitable for further manufacturing

Synonym(s):

Polymerase, Reverse Transcriptase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
eCl@ss:
32035490
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

form

liquid

storage condition

avoid repeated freeze/thaw cycles

technique(s)

RT-qPCR: suitable (Isothermal amplification), cDNA synthesis: suitable

impurities

(DNase free, RNasefree, DNA free, NICKase free)

color

colorless

suitability

suitable for PCR (Isothermal amplification)

storage temp.

−20°C (−15°C to −25°C)

General description

BST-RT Polymerase is an engineered Bst enzyme with robust reverse transcriptase (RT) activity, making it ideal for use in loop-mediated isothermal amplification (LAMP) or RT-LAMP assays. This feature eliminates the necessity of a secondary RT enzyme when targeting amplification from RNA samples, simplifying reaction setup. BST-RT works optimally at 65°C and can be used for real time reaction monitoring with a fluorescent dye or in endpoint detection assays. BST-RT polymerase is tolerant to potential reaction inhibitors such as ethanol, extraction buffers, and saliva. This enzyme is also compatible with workflows that incorporate dUTP for elimination of reaction carryover contaminants.

Application

Loop-mediated isothermal amplification (LAMP) has emerged as a key alternative to polymerase chain reaction (PCR)-based techniques for amplifying nucleic acids. While conventional PCR employs a thermal cycler for repetitive cycling temperatures when amplifying, isothermal amplification techniques occur at a single and fixed temperature, allowing the use of simple, portable, and more robust instruments for fast and exponential amplification.

Features and Benefits

  • Compatible with samples containing either DNA orRNA
  • Ability to perform RT-LAMP without additional RTenzymes
  • Lacks 5′ to 3′ exonuclease domain
  • DNase free, RNase free, DNA free, and Nickase free
  • Active from 25-65 °C (optimal at 65 °C)
  • Unit Definition: One unit is the amount of enzyme required to incorporate 10 nmol of dNTP onto ssDNA template for 30minutes at 65°C
  • Also available in stand-alone, high concentrations and glycerol-free formulations



Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

7783-20-2

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library