Merck
  • Amino acid analysis: aqueous dimethyl sulfoxide as solvent for the ninhydrin reaction.
Amino acid analysis: aqueous dimethyl sulfoxide as solvent for the ninhydrin reaction.
The Journal of biological chemistry (1968-12-10)
S Moore
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Lithium acetate dihydrate, BioUltra, ≥99.0% (anhydrous basis)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ninhydrin Reagent, 2% Solution
Sigma-Aldrich
Ninhydrin, ACS reagent