Merck
  • Differences in Mitotic Spindle Architecture in Mammalian Neural Stem Cells Influence Mitotic Accuracy during Brain Development.
Differences in Mitotic Spindle Architecture in Mammalian Neural Stem Cells Influence Mitotic Accuracy during Brain Development.
Current biology : CB (2019-09-10)
Diana Vargas-Hurtado, Jean-Baptiste Brault, Tristan Piolot, Ludovic Leconte, Nathalie Da Silva, Carole Pennetier, Alexandre Baffet, Véronique Marthiens, Renata Basto
ABSTRACT
A functional bipolar spindle is essential to segregate chromosomes correctly during mitosis. Across organisms and cell types, spindle architecture should be optimized to promote error-free divisions. However, it remains to be investigated whether mitotic spindle morphology adapts to changes in tissue properties, typical of embryonic development, in order to ensure different tasks, such as spindle positioning and chromosome segregation. We have characterized mitotic spindles in neural stem cells (NSCs) of the embryonic developing mouse neocortex. Surprisingly, we found a switch in spindle morphology from early to late neurogenic stages, which relies on an increase in inner spindle microtubule density and stability. Mechanistically, we identified the microtubule-associated protein TPX2 as one determinant of spindle shape, contributing not only to its robustness but also to correct chromosome segregation upon mitotic challenge. Our findings highlight a possible causal relationship between spindle architecture and mitotic accuracy with likely implications in brain size regulation.
Product Description
Supelco
Fast Green FCF, analytical standard
Sigma-Aldrich
Monoclonal Anti-β-Tubulin III antibody produced in mouse, clone SDL.3D10, ascites fluid
Sigma-Aldrich
Poly-D-lysine hydrobromide, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O, Molecular Biology
Sigma-Aldrich
Anti-Tubulin Antibody, Detyrosinated, Chemicon®, from rabbit
Sigma-Aldrich
Anti-PAX6 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, purified from hybridoma cell culture
Sigma-Aldrich
hBFGF, FGF-Basic, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%