Merck
  • Simultaneous detection of the enzyme activities of GPx1 and GPx4 guide optimization of selenium in cell biological experiments.
Simultaneous detection of the enzyme activities of GPx1 and GPx4 guide optimization of selenium in cell biological experiments.
Redox biology (2020-04-12)
Jeffrey M Stolwijk, Kelly C Falls-Hubert, Charles C Searby, Brett A Wagner, Garry R Buettner
ABSTRACT
Selenium is a metalloid trace element essential for maintaining the optimal redox environment in cells and tissues. It is structurally incorporated into over 25 selenoproteins and enzymes. The glutathione peroxidase (GPx) family of enzymes has a critical role in human health because of its antioxidant function. The recommended daily allowance (RDA) for selenium intake in humans was established to maximize the activity of GPx in plasma. Suboptimal availability of selenium can limit the expression and activities of GPxs leading to a compromised redox environment. This can cause detrimental oxidative distress that could be prevented by increasing the availability of selenium. In cell culture studies, the medium is typically deficient in selenium; supplementation with selenium can increase selenoenzyme activities. However, the optimal level of supplementation in cell culture media has not been well characterized. We performed dose-response experiments for the activities of GPx1 and GPx4 vs. the level of selenium supplementation in cell culture medium. For this, we advanced an assay to determine the activities of both GPx1 and GPx4 efficiently in a single run. During the optimization process, we found that the observed activities of GPx1 and GPx4 depend greatly on the pH of the assay buffer; the observed activities increase with increasing pH, with pH 8 being optimal. Using the combination assay, we also found that the expression and activities for both GPx1 and GPx4 can be maximized in exponentially growing cells by supplementing cell culture media with ≈ 200 nM seleno-l-methionine, without concerns for toxicity. Optimizing the availability of selenium in cell culture to maximize the expression and activities GPx1 and GPx4 may allow for better translation of information from cell culture work to in vivo settings.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt hydrate, ≥97% (HPLC)
Sigma-Aldrich
L-α-Phosphatidylcholine, solution, from soybean, suitable for cell culture, Type III-S
Sigma-Aldrich
Seleno-L-methionine, ≥98% (TLC), powder
Sigma-Aldrich
Phenazine methosulfate
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
1-Methoxy-5-methylphenazinium methyl sulfate, ≥95%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Lipoxidase from Glycine max (soybean), Type V, ammonium sulfate suspension, 500,000-1,000,000 units/mg protein
Sigma-Aldrich
Glutathione Reductase from baker′s yeast (S. cerevisiae), ammonium sulfate suspension, 100-300 units/mg protein (biuret)
Sigma-Aldrich
Triton X-100, peroxide- and carbonyl-free
Sigma-Aldrich
Thioredoxin Reductase Assay Kit, 1 kit sufficient for 100 assays (1 mL)
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse