Merck
  • Lamin A-dependent misregulation of adult stem cells associated with accelerated ageing.
Lamin A-dependent misregulation of adult stem cells associated with accelerated ageing.
Nature cell biology (2008-03-04)
Paola Scaffidi, Tom Misteli
ABSTRACT
The premature-ageing disease Hutchinson-Gilford Progeria Syndrome (HGPS) is caused by constitutive production of progerin, a mutant form of the nuclear architectural protein lamin A. Progerin is also expressed sporadically in wild-type cells and has been linked to physiological ageing. Cells from HGPS patients exhibit extensive nuclear defects, including abnormal chromatin structure and increased DNA damage. At the organismal level, HGPS affects several tissues, particularly those of mesenchymal origin. How the cellular defects of HGPS cells lead to the organismal defects has been unclear. Here, we provide evidence that progerin interferes with the function of human mesenchymal stem cells (hMSCs). We find that expression of progerin activates major downstream effectors of the Notch signalling pathway. Induction of progerin in hMSCs changes their molecular identity and differentiation potential. Our results support a model in which accelerated ageing in HGPS patients, and possibly also physiological ageing, is the result of adult stem cell dysfunction and progressive deterioration of tissue functions.
Product Description
Supelco
Oil Red O, BioReagent, suitable for electrophoresis
Sigma-Aldrich
Oil Red O, certified by the BSC
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
γ Secretase Inhibitor, ≥95% (HPLC)
Sigma-Aldrich
Alcian Blue 8GX, certified by the BSC
Sigma-Aldrich
Alcian Blue 8GX, powder
Sigma-Aldrich
Alizarin Red S, certified by the BSC
Sigma-Aldrich
β-Secretase inhibitor, ≥97% (HPLC), powder
Sigma-Aldrich
Alcian Blue 8GX, BioReagent, suitable for detection of glycoproteins
Sigma-Aldrich
Alcian Blue 8GX