Merck
  • The mych gene is required for neural crest survival during zebrafish development.
The mych gene is required for neural crest survival during zebrafish development.
PloS one (2008-05-01)
Sung-Kook Hong, Michael Tsang, Igor B Dawid
ABSTRACT
Among Myc family genes, c-Myc is known to have a role in neural crest specification in Xenopus and in craniofacial development in the mouse. There is no information on the function of other Myc genes in neural crest development, or about any developmental role of zebrafish Myc genes. We isolated the zebrafish mych (myc homologue) gene. Knockdown of mych leads to severe defects in craniofacial development and in certain other tissues including the eye. These phenotypes appear to be caused by cell death in the neural crest and in the eye field in the anterior brain. Mych is a novel factor required for neural crest cell survival in zebrafish.
Sigma-Aldrich
Alcian Blue 8GX
Sigma-Aldrich
Alcian Blue 8GX, powder
Sigma-Aldrich
Alcian Blue 8GX, certified by the BSC
Sigma-Aldrich
Alcian Blue 8GX, BioReagent, suitable for detection of glycoproteins
Sigma-Aldrich
Methyl Green, zinc chloride salt, ~85%
Sigma-Aldrich
Methyl Green, zinc chloride salt, certified by the BSC, Dye content 85 %
Sigma-Aldrich
Methyl Green, zinc chloride salt, <0.5% crystal violet, Dye content 80 %
Sigma-Aldrich
Methyl Green, zinc chloride salt, suitable for microscopy