Merck
  • Effectors Enabling Adaptation to Mitochondrial Complex I Loss in Hürthle Cell Carcinoma.
Effectors Enabling Adaptation to Mitochondrial Complex I Loss in Hürthle Cell Carcinoma.
Cancer discovery (2023-06-01)
Raj K Gopal, Venkata R Vantaku, Apekshya Panda, Bryn Reimer, Sneha Rath, Tsz-Leung To, Adam S Fisch, Murat Cetinbas, Maia Livneh, Michael J Calcaterra, Benjamin J Gigliotti, Kerry A Pierce, Clary B Clish, Dora Dias-Santagata, Peter M Sadow, Lori J Wirth, Gilbert H Daniels, Ruslan I Sadreyev, Sarah E Calvo, Sareh Parangi, Vamsi K Mootha
ABSTRACT
Oncocytic (Hürthle cell) carcinoma of the thyroid (HCC) is genetically characterized by complex I mitochondrial DNA mutations and widespread chromosomal losses. Here, we utilize RNA sequencing and metabolomics to identify candidate molecular effectors activated by these genetic drivers. We find glutathione biosynthesis, amino acid metabolism, mitochondrial unfolded protein response, and lipid peroxide scavenging to be increased in HCC. A CRISPR-Cas9 knockout screen in a new HCC model reveals which pathways are key for fitness, and highlights loss of GPX4, a defense against lipid peroxides and ferroptosis, as a strong liability. Rescuing complex I redox activity with the yeast NADH dehydrogenase (NDI1) in HCC cells diminishes ferroptosis sensitivity, while inhibiting complex I in normal thyroid cells augments ferroptosis induction. Our work demonstrates unmitigated lipid peroxide stress to be an HCC vulnerability that is mechanistically coupled to the genetic loss of mitochondrial complex I activity. HCC harbors abundant mitochondria, mitochondrial DNA mutations, and chromosomal losses. Using a CRISPR-Cas9 screen inspired by transcriptomic and metabolomic profiling, we identify molecular effectors essential for cell fitness. We uncover lipid peroxide stress as a vulnerability coupled to mitochondrial complex I loss in HCC. See related article by Frank et al., p. 1884. This article is highlighted in the In This Issue feature, p. 1749.
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
1S,3R-RSL 3, ≥98% (HPLC)
Sigma-Aldrich
Ferrostatin-1, ≥95% (HPLC)
Sigma-Aldrich
Anti-HNE Fluorophore Rabbit pAb, liquid, Calbiochem®