Merck
  • Purification and characterization of an alkaline cellulase produced by Bacillus subtilis (AS3).
Purification and characterization of an alkaline cellulase produced by Bacillus subtilis (AS3).
Preparative biochemistry & biotechnology (2013-02-06)
Deepmoni Deka, Mohammad Jawed, Arun Goyal
ABSTRACT
An extracellular alkaline carboxymethycellulase (CMCase) from Bacillus subtilis was purified by salt precipitation followed by anion-exchange chromatography using DEAE-Sepharose. The cell-free supernatant containing crude enzyme had a CMCase activity of 0.34 U/mg. The purified enzyme gave a specific activity of 3.33 U/mg, with 10-fold purification and an overall activity yield of 5.6%. The purified enzyme displayed a protein band on sodium dodecyl sulfate polyacrylamide gel electrophoresis (SDS-PAGE) with an apparent molecular size of 30 kDa, which was also confirmed by zymogram analysis. The enzyme displayed multisubstrate specificity, showing significantly higher activity with lichenan and β-glucan as compared to carboxymethylcellulose (CMC), laminarin, hydroxyethylcellulose, and steam-exploded bagasse, and negligible activity with crystalline substrate such as Avicel and filter paper. It was optimally active at pH 9.2 and temperature 45°C. The enzyme was stable in the pH range 6-10 and retained 70% activity at pH 12. Thermal stability analysis revealed that the enzyme was stable in temperature range of 20°C to 45°C and retained more than 50% activity at 60°C for 30 min. The enzyme had a Km of 0.13 mg/ml and Vmax of 3.38 U/mg using CMC as substrate.
Sigma-Aldrich
Hydroxyethyl-cellulose
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.9
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.7
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 1.2
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 2500-6000 cP 
Sigma-Aldrich
Carboxymethylcellulose sodium, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
Hydroxyethyl-cellulose, viscosity 90-160 cP, 5 % in H2O(25 °C)
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 50-200 cP , c=4% H2O at 25­°C
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mw ~380,000
Sigma-Aldrich
Laminarin from Laminaria digitata, polysaccharide substrate for laminarinase
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, low viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mv ~90,000
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mv ~720,000
Sigma-Aldrich
2-Hydroxyethyl cellulose, average Mv ~1,300,000
Sigma-Aldrich
Carboxymethylcellulose sodium salt, medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, ultra high viscosity, highly purified