Merck
  • Effects of oral administration of zinc and diiodohydroxyquinolein on plasma zinc levels of uremic patients.
Effects of oral administration of zinc and diiodohydroxyquinolein on plasma zinc levels of uremic patients.
Nephron (1995-01-01)
R Paniagua, R Claure, D Amato, E Flores, A Pérez, E Exaire
ABSTRACT
Patients with chronic renal failure often have low plasma zinc (Zn) levels. Some factors that may account for abnormal Zn metabolism in these patients are low dietary Zn intake, a specific Zn transport defect, or absence of intestinal Zn ligand. In this study Zn supplementation and a Zn-chelating drug, diiodohydroxyquinolein (DQ), were used to assess the effects of Zn intake and Zn transporters on Zn plasma levels in patients with chronic renal failure. To meet this objective, 20 uremic patients were randomly assigned to one of the following groups of treatment: group 1 received placebo; group 2 Zn sulfate (100 mg/day p.o.), group 3 DQ (80 mg/day p.o.), and group 4 received Zn sulfate plus DQ at the same dosages as in groups 2 and 3. The Zn plasma levels were measured in venous samples, before and after 1 and 2 weeks of treatment, by atomic absorption spectrophotometry. The Zn plasma levels increased in group 2 patients from 8 +/- 0.2 to 10 +/- 0.4 and 11 +/- 0.9 mumol/l by the end of the 1st and 2nd weeks of treatment, respectively. In group 4 patients, the Zn plasma levels increased even more: from 9 +/- 0.1 to 14 +/- 1.6 and 13 +/- 2.1 mumol/l respectively. The plasma Zn concentration of group 1 and 3 patients remained at basal levels. These results show that DQ, when given along with Zn sulfate supplements, causes a greater increase in plasma Zn levels than that caused by either drug given alone.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
L-2-Aminobutyric acid, ≥99% (titration)
Sigma-Aldrich
L-2-Aminobutyric acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Trimethylamine N-oxide, 95%
Sigma-Aldrich
Isomaltose, ~98% (TLC)
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
1-Methyl-L-histidine, ≥98.0% (TLC)
Sigma-Aldrich
4-Heptanone, 98%
Supelco
4-Heptanone, analytical standard
Sigma-Aldrich
3-Methyl-L-histidine
Sigma-Aldrich
3-Hydroxymandelic acid, ≥97.0% (T)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Zinc, powder, <150 μm, 99.995% trace metals basis
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Zinc, purum, powder
Sigma-Aldrich
Zinc, granular, 30-100 mesh, 99%
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Dimethylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Dimethylamine solution, 2.0 M in methanol
Sigma-Aldrich
Dimethylamine solution, 2.0 M in THF
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol