Merck
  • Ethanol, Zn2+ and insulin interact as progression factors to enhance DNA synthesis synergistically in the presence of Ca2+ and other cell cycle initiators in fibroblasts.
Ethanol, Zn2+ and insulin interact as progression factors to enhance DNA synthesis synergistically in the presence of Ca2+ and other cell cycle initiators in fibroblasts.
The Biochemical journal (2000-02-05)
J S Huang, Q B She, K S Crilly, Z Kiss
ABSTRACT
In serum-starved NIH 3T3 fibroblasts, ethanol (30-80 mM) promoted the effects of insulin and insulin-like growth factor I (IGF-I) on DNA synthesis in a Zn(2+)-dependent manner. Ethanol and Zn(2+) were most effective when added shortly before or after insulin, indicating that all these agents facilitated cell cycle progression. The synergistic effects of ethanol, Zn(2+) and insulin (or IGF-I) on DNA synthesis required 1.1-2.3 mM Ca(2+), which seemed to act as the cell cycle initiator. When serum-starved cells were pretreated for 2 h with other cell cycle initiators such as 10% (v/v) serum, 50 ng/ml platelet-derived growth factor or 2 ng/ml fibroblast growth factor, subsequent co-treatments with 60 mM ethanol, Zn(2+) and insulin for an 18 h period again synergistically increased DNA synthesis. Of the various signal transducing events examined, ethanol stimulated cellular uptake of (45)Ca and it enhanced the stimulatory effects of insulin on p70 S6 kinase activity in a Zn(2+)-dependent manner. In contrast, ethanol inhibited insulin-induced activating phosphorylation of p42/p44 mitogen-activated protein kinases; these inhibitory ethanol effects were prevented by Zn(2+). The results show that, in NIH 3T3 fibroblasts, ethanol can promote cell cycle progression in the presence of a cell cycle initiator as well as Zn(2+) and insulin (or IGF-I).
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Ethanol
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)