Merck
  • Cerebrospinal fluid levels of thiamine in patients with Alzheimer's disease.
Cerebrospinal fluid levels of thiamine in patients with Alzheimer's disease.
Journal of neural transmission (Vienna, Austria : 1996) (2002-07-12)
J A Molina, F J Jiménez-Jiménez, A Hernánz, E Fernández-Vivancos, S Medina, F de Bustos, C Gómez-Escalonilla, Y Sayed
ABSTRACT
Thiamine is an essential cofactor for several important enzymes involved in brain oxidative metabolism, such as the alpha-ketoglutarate dehydrogenase complex (KGDHC), pyruvate-dehydrogenase complex (PDHC), and transketolase. Some investigators reported decreased thiamine-diphosphate levels and decreased activities of KGDHC, pyruvate-dehydrogenase complex and transketolase in the brain tissue of Alzheimer's disease (AD) patients. We measured cerebrospinal (CSF) levels of thiamine-diphosphate, thiamine-monophosphate, free thiamine, and total thiamine, using ion-pair reversed phase high performance liquid chromatography, in 33 patients with sporadic AD and 32 matched controls. The mean CSF levels of thiamine-derivatives did not differ significantly from those of controls, while the mean plasma levels of thiamine-diphosphate, free and total thiamine were significantly lower in the AD-patient group. CSF and plasma thiamine levels were not correlated with age, age at onset, duration of the disease, and scores of the MiniMental State Examination, with the exception of plasma thiamine-diphosphate with MiniMental State Examination (r = 0.41, p < 0.05) in the AD-patients group. CSF and plasma values did not predict dementia progression, assessed with the MiniMental State Examination scores. These results suggest that CSF thiamine levels are not related with the risk for and the progression of AD.
Product Description
Supelco
L-Phenylalanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Methylmalonic acid, 99%
Sigma-Aldrich
Copper, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Asparagine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Silicon, pieces, 99.95% trace metals basis
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Silicon, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Supelco
Fumaric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Titanium, IRMM®, certified reference material, 0.1 mm foil
SAFC
L-Isoleucine
Sigma-Aldrich
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Succinic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Tungsten, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.9% trace metals basis