Merck
  • Sulpho-conjugation of ethanol in humans in vivo and by individual sulphotransferase forms in vitro.
Sulpho-conjugation of ethanol in humans in vivo and by individual sulphotransferase forms in vitro.
The Biochemical journal (2004-07-30)
Heiko Schneider, Hansruedi Glatt
ABSTRACT
We studied whether ethanol is sulphonated in humans with the perspective of using the urinary excretion of ethyl sulphate after ethanol consumption as a biomarker for SULT (sulphotransferase) activity. We developed a sensitive and selective HPLC-MS/MS method for determining ethyl sulphate in urine. Ten volunteers received a low dose of ethanol (0.1 g/kg of body mass). In general, excretion of ethyl sulphate was maximal in the first or second hour after dosage. Within 8 h, 2.5-6.8 micromol of ethyl sulphate was excreted. A 5-fold increase in the dose of ethanol led to an increase in the amount of ethyl sulphate excreted within 8 h (28-95 micromol) and the presence of this metabolite in urine for at least 24 h. Since ethyl sulphate was still being excreted for a substantial period after the elimination of ethanol, it might be used as a medium-time biomarker for preceding ethanol consumption. We have expressed previously all human SULT forms identified in Salmonella typhimurium. Ethanol sulphonation was studied in cytosolic preparations of these strains. The highest activities were observed with SULT1A2, 1B1 and 1C2, followed by 1A3. Activities were markedly lower with SULT1E1, 1A1 and 2A1, and were negligible with SULT1C1, 2B1a, 2B1b and 4A1. If the expression levels in tissues are additionally taken into account, SULT1A3 might be the predominant form for the sulphonation of ethanol in vivo, although a robust estimate requires further studies. With this limitation, urinary ethyl sulphate excretion appears very promising as a biomarker for SULT activity in vivo.
Product Description
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Hexanal, 98%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Hexanal, ≥97%, FCC, stabilized, FG
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Hexanal, natural, ≥95%, FG
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)