Merck
  • A single bout of exercise is followed by a prolonged decrease in the interstitial glucose concentration in skeletal muscle.
A single bout of exercise is followed by a prolonged decrease in the interstitial glucose concentration in skeletal muscle.
Acta physiologica Scandinavica (2005-11-04)
J Henriksson, M Knol
ABSTRACT
The present study was performed to test the hypothesis that the interstitial glucose concentration in human skeletal muscle is decreased for a prolonged period following a single bout of exercise, while blood flow has returned to resting levels. Muscle interstitial concentrations of glucose, lactate, pyruvate and urea were monitored in six healthy individuals during 8 h following a 2-h one-leg exercise session by microdialysis at low perfusion flow rate. Simultaneously the blood flow was measured by the microdialysis ethanol technique. The blood glucose and the control leg interstitial glucose concentrations were stable during the experiment averaging 5.7 +/- 0.1 and 4.1 +/- 0.3 mm, respectively. In contrast, the interstitial glucose concentration in the exercise leg was markedly decreased, averaging 1.9 +/- 0.5 mm, during the first 5.5 h following exercise (P < 0.01), after which it returned towards normal values. Muscle blood flow at the site of the microdialysis catheter, measured as the ethanol outflow-to-inflow ratio, did not change significantly over time in the control or exercise leg and did not differ significantly between the two legs. Interstitial concentrations of lactate, pyruvate and urea were not significantly different between the control and exercise leg. The study shows that the interstitial glucose concentration in skeletal muscle is markedly decreased for several hours following a single exercise session. The decreased interstitial glucose concentration may serve to limit the rate of post-exercise muscle glucose uptake to a rate compatible with normal blood glucose levels and may also be speculated to have a positive long-term health implication by augmenting muscle insulin sensitivity.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology