Merck
  • Hemodynamic and metabolic responses to interstitial angiotensin II in normal weight and obese men.
Hemodynamic and metabolic responses to interstitial angiotensin II in normal weight and obese men.
Journal of hypertension (2006-05-11)
Michael Boschmann, Frauke Adams, Karin Schaller, Gabriele Franke, Arya M Sharma, Susanne Klaus, Friedrich C Luft, Jens Jordan
ABSTRACT
The expression of the AT1 receptor in adipose tissue is not decreased or even increased in obese subjects despite systemic activation of the renin-angiotensin system. Therefore, we hypothesized that peripheral tissues of obese subjects are hypersensitive to angiotensin (Ang) II. We characterized the effect of locally applied Ang II in skeletal muscle and subcutaneous adipose tissue of healthy non-obese (n = 12) and obese (n = 11) men using the microdialysis technique. Tissues were perfused with Ringer's solution + ethanol and incremental doses of Ang II (0.01, 0.1 and 1 micromol/l). Dialysate ethanol, glycerol, glucose, lactate, and pyruvate concentrations were measured to assess changes in blood flow (ethanol dilution technique), lipolysis and glycolysis, respectively. In adipose tissue, basal ethanol ratio was significantly higher and dialysate metabolite concentrations were significantly lower in obese versus non-obese men. In muscle, basal dialysate glycerol was significantly higher in obese versus non-obese men. Ang II elicited small increases in ethanol ratio and decreases in dialysate glucose in adipose tissue and skeletal muscle in both non-obese and obese men. Dialysate lactate increased significantly in both tissues of obese, but not non-obese men. Dialysate glycerol increased in adipose tissue of non-obese (+ 40%) but not of obese and remained almost unchanged in muscle of both groups. Interstitially applied Ang II elicits subtle changes in tissue perfusion and metabolism. However, we did not find a major increase in interstitial Ang II responsiveness in obese men.
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)