Merck
Influence of alcohol on peripheral neurotransmitter function.
Federation proceedings (1982-06-01)
L A Pohorecky
ABSTRACT
Presently available evidence for the action of ethanol on the peripheral noradrenergic system is reviewed. Acute ethanol treatment produces an initial elevation in catecholamine levels in blood, and in the excretion of catecholamines, which may be followed by a decrease in catecholamine release. Correspondingly, a hyposensitivity of peripheral beta receptors has been noted. Other tissues such as vascular smooth muscles show a biphasic response to ethanol: a hypersensitivity to norepinephrine at low concentrations of ethanol, and hyposensitivity to norepinephrine at high ethanol concentrations. In the adrenal medulla the activity of various enzymes involved in catecholamine synthesis is increased by ethanol treatment. Chronic treatment with ethanol does not alter endogenous norepinephrine levels in heart, but does increase its turnover. Chronic treatment also lowers the number of beta-adrenergic receptors in the heart, and decreases the response to peripherally administered isoproterenol, which suggests a hyposensitivity of peripheral beta receptors. Tolerance may develop to the chronic effects of ethanol in the heart and the vas deferens, but in the adrenals the activity of enzymes for catecholamine synthesis remains elevated with chronic treatment. During withdrawal, there is generalized hyperexcitability of the sympathetic nervous system. Norepinephrine and amine turnover in the heart is increased, as is the activity of adrenal medullary enzymes involved in catecholamine synthesis.
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications