Merck
  • Transdermal nitroglycerin for the treatment of preterm labor: a systematic review and metaanalysis.
Transdermal nitroglycerin for the treatment of preterm labor: a systematic review and metaanalysis.
American journal of obstetrics and gynecology (2013-07-31)
Agustín Conde-Agudelo, Roberto Romero
ABSTRACT
The purpose of this study was to evaluate the efficacy and safety of transdermal nitroglycerin as a tocolytic agent in women with preterm labor. We conducted a systematic review and metaanalysis of randomized controlled trials. Thirteen studies were included (1302 women) comparing transdermal nitroglycerin vs placebo (2 studies; n = 186); β2-adrenergic receptor agonists (9 studies; n = 1024); nifedipine (1 study; n = 50); and magnesium sulfate (1 study; n = 42). There were no significant differences between transdermal nitroglycerin and placebo for delivery within 48 hours of the initiation of treatment or at <28, <34, or <37 weeks of gestation, adverse neonatal outcomes, and neurodevelopmental status at 24 months of life. Nevertheless, 1 study found a marginally significant reduction in the risk of a composite outcome of major neonatal morbidity and perinatal death (3/74 [4.1%] vs 11/79 [13.9%]; relative risk, 0.29; 95% confidence interval, 0.08-1.00). When compared with β2-adrenergic receptor agonists, transdermal nitroglycerin was associated with a significant reduction in the risk of preterm birth at <34 and <37 weeks of gestation, admission to the neonatal intensive care unit, use of mechanical ventilation, and maternal side effects. There were no significant differences between transdermal nitroglycerin and nifedipine and magnesium sulfate in delivery within 48 hours of treatment and pregnancy prolongation, respectively. Overall, women who received transdermal nitroglycerin had a higher risk of headache. Although transdermal nitroglycerin appears to be more effective than β2-adrenergic receptor agonists, the current evidence does not support its routine use as a tocolytic agent for the treatment of preterm labor. Further double-blind placebo-controlled trials are needed.
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Nifedipine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium sulfate heptahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Magnesium sulfate heptahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioUltra, ≥99.5% (KT)
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioReagent, Molecular Biology, suitable for plant cell culture, ≥99.0%
Supelco
Trinitroglycerin solution, 1 % (w/w) in propylene glycol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioXtra, ≥99.0%