Merck
  • Utilization and safety of sodium hyaluronate-carboxymethylcellulose adhesion barrier.
Utilization and safety of sodium hyaluronate-carboxymethylcellulose adhesion barrier.
Diseases of the colon and rectum (2013-09-12)
Shazia Bashir, Cande V Ananth, Sharyn N Lewin, William M Burke, Yu-Shiang Lu, Alfred I Neugut, Thomas J Herzog, Dawn L Hershman, Jason D Wright
ABSTRACT
Little is known about the use and toxicity of antiadhesion substances such as sodium hyaluronate-carboxymethylcellulose. We analyzed the patterns of use and safety of sodium hyaluronate-carboxymethylcellulose in patients undergoing colectomy and gynecologic surgery. This is a retrospective cohort study. This study covered hospitals nationwide. All patients in the Premier Perspective database who underwent colectomy or hysterectomy from 2000 to 2010 were included in the analyses. Hyaluronate-carboxymethylcellulose use was determined by billing codes. For the primary outcome, we used hierarchical mixed-effects logistic regression models to determine the factors associated with the use of hyaluronate-carboxymethylcellulose, whereas a propensity score-matched analysis was used to secondarily assess the association between hyaluronate-carboxymethylcellulose use and toxicity (abscess, bowel and wound complications, peritonitis). We identified 382,355 patients who underwent hysterectomy and 267,368 who underwent colectomy. For hysterectomy, hyaluronate-carboxymethylcellulose use was 5.0% overall, increasing from 1.1% in 2000 to 9.8% in 2010. Hyaluronate-carboxymethylcellulose was used in 8.1% of those who underwent colectomy and increased from 6.2% in 2000 to 12.4% in 2010. The year of diagnosis and procedure volume of the attending surgeon were the strongest predictors of hyaluronate-carboxymethylcellulose use. After matching and risk adjustment, hyaluronate-carboxymethylcellulose use was not associated with abscess formation (1.5% vs 1.5%) (relative risk = 0.97; 95% CI, 0.84-1.12) in those who underwent hysterectomy. A patient receiving hyaluronate-carboxymethylcellulose had a 13% increased risk of abscess (17.4% vs 15.0%) (relative risk = 1.13; 95% CI, 1.08-1.17) after colectomy. This was an observational study. Hyaluronate-carboxymethylcellulose use has increased over the past decade for colectomy and hysterectomy. Although there is no association between hyaluronate-carboxymethylcellulose use and abscess following hysterectomy, hyaluronate-carboxymethylcellulose use was associated with a small increased risk of abscess after colectomy.
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.9
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 50-200 cP , c=4% H2O at 25­°C
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.7
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 1.2
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 2500-6000 cP 
Sigma-Aldrich
Carboxymethylcellulose sodium, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, low viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, ultra high viscosity, highly purified
Sigma-Aldrich
Carboxymethylcellulose sodium salt, medium viscosity