Merck
  • Quantitative characterization of the inhibitory effects of salt, humic acid, and heavy metals on the recovery of waterborne norovirus by electropositive filters.
Quantitative characterization of the inhibitory effects of salt, humic acid, and heavy metals on the recovery of waterborne norovirus by electropositive filters.
Journal of water and health (2013-12-18)
MinJung Kim, GwangPyo Ko
ABSTRACT
The virus adsorption-elution technique (VIRADEL) using electropositively charged filters is used frequently for recovering enteric viruses from water. The filter-absorbed virus is typically eluted, concentrated, and subsequently detected by culture or molecular methods. Human norovirus (HuNoV), one of the most important waterborne pathogens, cannot be cultivated by conventional culture methods and is typically detected using a reverse transcriptase-polymerase chain reaction (RT-PCR) assay. However, it is plausible that various inhibitors could be concentrated simultaneously during the VIRADEL process and affect RT-PCR assays. In this study, we evaluated the effect of typical inhibitors, including humic acid, heavy metals, and salt, on the recovery of norovirus by two different electropositive filters: 1MDS and Nanoceram. Known amounts of HuNoV and murine norovirus were inoculated in 1 L of surface water containing various concentrations of humic acid, heavy metals (cadmium and lead), or NaCl. Our results indicate that the presence of heavy metals or salt significantly reduced the recovery of virus from the electropositive filters. Thus, care should be taken when analyzing waterborne norovirus using electropositive filters in environments with high concentrations of heavy metal inhibitors or salts.
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Lead, powder, −325 mesh, ≥99% trace metals basis
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis