Merck
The evaluation of the amebicidal activity of Eryngium planum extracts.
Acta poloniae pharmaceutica (2014-01-05)
Monika Derda, Barbara Thiem, Jaromir Budzianowski, Waldemar Jerzy Wojt, Agnieszka Wojtkowiak-Giera
ABSTRACT
Selected fractions of ethanolic extracts obtained from leaves and roots of Eryngium planum (Apiaceae) were evaluated in vitro for amebicidal activity against Acanthamoeba castellanii. This free-living ameba is the cause of Acanthamoeba keratitis, which is a painful, vision-impairing disease of the eyes, and chronic granulomatous amebic encephalitis. Treatment is very difficult and not always effective because of encystation, which makes the amebae highly resistant to anti-amebic drugs. The search for novel natural amebicidal agents is still of current interest. Fractions of E. planum ethanolic extract from basal leaves: flavonoid fraction (Lf), flavonoid-saponin fraction (Lf-s), saponin fraction (Ls) and phenolic acids fraction (La) and from roots: saponin fraction (Rs) and phenolic acids fraction (Ra) were assayed for antiamebic activity. In the presence of the saponin fractions and phenolic acid fractions (ranging from 1-5 mg/mL), the number of the trofozoites of Acanthamoeba castellanii viable strain 309 decreased during the experimental period (0-72 h). On the other hand, the flavonoid fraction from leaves showed a stimulating activity on the amebae. Almost all fractions (except the flavonoid fraction) showed a time- and dose-dependent amebistatic activity on the trophozoites. Of the fractions tested, the phenolic acid fraction from roots at the concentration of 5 mg/L showed the amebicidal activity on the trophozoites.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%