Merck
  • Effects of combined exogenous dextranase and sodium fluoride on Streptococcus mutans 25175 monospecies biofilms.
Effects of combined exogenous dextranase and sodium fluoride on Streptococcus mutans 25175 monospecies biofilms.
American journal of dentistry (2014-02-01)
Ying-Ming Yang, Dan Jiang, Yuan-Xin Qiu, Rong Fan, Ru Zhang, Mei-Zhi Ning, Mei-Ying Shao, Chao-Liang Zhang, Xiao Hong, Tao Hu
ABSTRACT
To investigate the effects of exogenous dextranase and sodium fluoride on a S. mutans monospecies biofilm. S. mutans 25175 was grown in tryptone soya broth medium, and biofilm was formed on glass slides with 1.0% sucrose. Exogenous dextranase and sodium fluoride were added alone or together. The biofilm morphology was analyzed by confocal laser scanning microscopy. The effects of the drug on the adhesion and exopolysaccharide production by the biofilms were evaluated by scintillation counting and the anthrone method, respectively. In this study, we found that the structure of initial biofilm and mature biofilm were partly altered by dextranase and high concentrations of sodium fluoride separately. However, dextranase combined with a low concentration of sodium fluoride could clearly destroy the typical tree-like structure of the biofilm, and led to less bacterial adhesion than when the dextranase or fluoride were used alone (P < 0.05). The amounts of soluble and insoluble exopolysaccharide were significantly reduced by combining dextranase with a low concentration of sodium fluoride, much more than when they were used alone (P < 0.05). These data indicate that dextranase and a low concentration of sodium fluoride may have synergistic effects against S. mutans biofilm and suggest the application of a low concentration of sodium fluoride in anticaries treatment.
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Dextranase from Penicillium sp., lyophilized powder, 10-25 units/mg solid
Sigma-Aldrich
Dextranase from Penicillium sp., lyophilized powder, 100-250 units/mg protein
Sigma-Aldrich
Dextranase from Penicillium sp., lyophilized powder, 400-800 units/mg protein
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sodium fluoride, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sustine 470
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium Fluoride Solution