Merck
  • Small-volume 7.5% NaCl adenosine, lidocaine, and Mg2+ has multiple benefits during hypotensive and blood resuscitation in the pig following severe blood loss: rat to pig translation.
Small-volume 7.5% NaCl adenosine, lidocaine, and Mg2+ has multiple benefits during hypotensive and blood resuscitation in the pig following severe blood loss: rat to pig translation.
Critical care medicine (2014-02-22)
Asger Granfeldt, Hayley L Letson, Janus A Hyldebrandt, Edward R Wang, Pablo A Salcedo, Torben K Nielsen, Else Tønnesen, Jakob Vinten-Johansen, Geoffrey P Dobson
ABSTRACT
Currently, there is no effective small-volume fluid for traumatic hemorrhagic shock. Our objective was to translate small-volume 7.5% NaCl adenosine, lidocaine, and Mg hypotensive fluid resuscitation from the rat to the pig. Pigs (35-40 kg) were anesthetized and bled to mean arterial pressure of 35-40 mm Hg for 90 minutes, followed by 60 minutes of hypotensive resuscitation and infusion of shed blood. Data were collected continuously. University hospital laboratory. Female farm-bred pigs. Pigs were randomly assigned to a single IV bolus of 4 mL/kg 7.5% NaCl + adenosine, lidocaine and Mg (n = 8) or 4 mL/kg 7.5% NaCl (n = 8) at hypotensive resuscitation and 0.9% NaCl ± adenosine and lidocaine at infusion of shed blood. At 60 minutes of hypotensive resuscitation, treatment with 7.5% NaCl + adenosine, lidocaine, and Mg generated significantly higher mean arterial pressure (48 mm Hg [95% CI, 44-52] vs 33 mm Hg [95% CI, 30-36], p < 0.0001), cardiac index (76 mL/min/kg [95% CI, 63-91] vs 47 mL/min/kg [95% CI, 39-57], p = 0.002), and oxygen delivery (7.6 mL O2/min/kg [95% CI, 6.4-9.0] vs 5.2 mL O2/min/kg [95% CI, 4.4-6.2], p = 0.003) when compared with controls. Pigs that received adenosine, lidocaine, and Mg/adenosine and lidocaine also had significantly lower blood lactate (7.1 mM [95% CI, 5.7-8.9] vs 11.3 mM [95% CI, 9.0-14.1], p = 0.004), core body temperature (39.3°C [95% CI, 39.0-39.5] vs 39.7°C [95% CI, 39.4-39.9]), and higher base excess (-5.9 mEq/L [95% CI, -8.0 to -3.8] vs -11.2 mEq/L [95% CI, -13.4 to -9.1]). One control died from cardiovascular collapse. Higher cardiac index in the adenosine, lidocaine, and Mg/adenosine and lidocaine group was due to a two-fold increase in stroke volume. Left ventricular systolic ejection times were significantly higher and inversely related to heart rate in the adenosine, lidocaine, and Mg/adenosine and lidocaine group. Thirty minutes after blood return, whole-body oxygen consumption decreased in pigs that received adenosine, lidocaine, and Mg/adenosine and lidocaine (5.7 mL O2/min/kg [95% CI, 4.7-6.8] to 4.9 mL O2/min/kg [95% CI, 4.2-5.8]), whereas it increased in controls (4.2 mL O2/min/kg [95% CI, 3.5-5.0] to 5.8 mL O2/min/kg [95% CI, 4.9-5.8], p = 0.02). After 180 minutes, pigs in the adenosine, lidocaine, and Mg/adenosine and lidocaine group had three-fold higher urinary output (2.1 mL//kg/hr [95% CI, 1.2-3.8] vs 0.7 mL//kg/hr [95% CI, 0.4-1.2], p = 0.001) and lower plasma creatinine levels. Small-volume resuscitation with 7.5% NaCl + adenosine, lidocaine, and Mg/adenosine and lidocaine provided superior cardiovascular, acid-base, metabolic, and renal recoveries following severe hemorrhagic shock in the pig compared with 7.5% NaCl alone.
Product Description
SAFC
Sodium chloride solution, 5 M
Lidocaine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Lidocaine, European Pharmacopoeia (EP) Reference Standard
Supelco
Lidocaine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis