Merck
  • Modulation of P-glycoprotein function and reversal of multidrug resistance by (-)-epigallocatechin gallate in human cancer cells.
Modulation of P-glycoprotein function and reversal of multidrug resistance by (-)-epigallocatechin gallate in human cancer cells.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2005-03-30)
Feng Qian, Dongzhi Wei, Qiang Zhang, Shengli Yang
ABSTRACT
Multidrug resistance (MDR) is a major obstacle in the chemotherapeutic treatment of many human cancers. In this study, the reversal of P-glycoprotein (P-gp) mediated multidrug resistance by (-)-epigallocatechin gallate (EGCG) and its molecular mechanism were investigated. A three-dimensional model of carboxyl-terminal nucleotide binding domain (NBD2) from P-gp was built by homology modeling. The structural model of the complex indicates that EGCG was tightly bound to the ATP-binding site of NBD2. EGCG modulated the function of P-gp and increased the intracellular accumulation of chemotherapeutic agent doxorubicin (DOX) in drug-resistant KB-A1 cells. When KB-A1 cells were exposed to 10 microg/ml DOX combined with 10, 30, 50 microM EGCG for 4 h, the intracellular concentrations of DOX were increased 1.5, 1.9, 2.3 times, respectively compared with DOX alone treatment. In vitro EGCG potentiated the cytotoxicity of DOX to drug-resistant KB-A1 cells. In KB-A1 cell xenograft model, EGCG could also enhance the efficacy of DOX and increased the DOX concentration in the resistant tumors. Thus, these results suggest that EGCG modulated the function of P-gp and reversed P-gp mediated multidrug resistance in human cancer cells.
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Epigallocatechin gallate, primary reference standard
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(−)-Epigallocatechin gallate, analytical standard