Merck
  • Identifying key non-volatile compounds in ready-to-drink green tea and their impact on taste profile.
Identifying key non-volatile compounds in ready-to-drink green tea and their impact on taste profile.
Food chemistry (2014-03-07)
Peigen Yu, Angelin Soo-Lee Yeo, Mei-Yin Low, Weibiao Zhou
ABSTRACT
Thirty-nine non-volatile compounds in seven ready-to-drink (RTD) green tea samples were analysed and quantified using liquid chromatography. Taste reconstruction experiments using thirteen selected compounds were conducted to identify the key non-volatile tastants. Taste profiles of the reconstructed samples did not differ significantly from the RTD tea samples. To investigate the taste contribution and significance of individual compounds, omission experiments were carried out by removing individual or a group of compounds. Sensory evaluation revealed that the astringent- and bitter-tasting (-)-epigallocatechin gallate, bitter-tasting caffeine, and the umami-tasting l-glutamic acid were the main contributors to the taste of RTD green tea. Subsequently, the taste profile of the reduced recombinant, comprising of a combination of these three compounds and l-theanine, was found to not differ significantly from the sample recombinant and RTD tea sample. Lastly, regression models were developed to objectively predict and assess the intensities of bitterness and astringency in RTD green teas.
Product Description
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Epicatechin, primary reference standard
Supelco
(−)-Epicatechin, analytical standard
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Supelco
Melting point standard 235-237°C, analytical standard
Supelco
(+)-Catechin, analytical standard
Supelco
(−)-Epigallocatechin gallate, analytical standard
Sigma-Aldrich
L-Glutamic acid, FCC
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Epigallocatechin gallate, primary reference standard
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, BioXtra
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Caffeine, European Pharmacopoeia (EP) Reference Standard
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg