Merck
  • Luteolin alleviates alcoholic liver disease induced by chronic and binge ethanol feeding in mice.
Luteolin alleviates alcoholic liver disease induced by chronic and binge ethanol feeding in mice.
The Journal of nutrition (2014-05-16)
Gaigai Liu, Yuxue Zhang, Chunchun Liu, Daqian Xu, Rui Zhang, Yuan Cheng, Yi Pan, Cheng Huang, Yan Chen
ABSTRACT
Ethanol consumption can lead to hepatic steatosis that contributes to late-stage liver diseases such as cirrhosis and hepatocellular carcinoma. In this study, we investigated the potential protective effect of a flavonoid, luteolin, on ethanol-induced fatty liver development and liver injury. Six-wk-old male C57BL/6 mice were divided into 3 groups: a control group; a group exposed to alcohol by using a chronic and binge ethanol feeding protocol (EtOH); and a group that was administered daily 50 mg/kg of luteolin in addition to ethanol exposure (EtOH + Lut). A chronic and binge ethanol feeding protocol was used, including chronic ethanol consumption (1%, 2%, and 4% for 3 d, and 5% for 9 d) and a binge (30% ethanol) on the last day. Compared with the control group, the EtOH group had a significant elevation in serum concentrations of alanine aminotransferase (ALT) (561%), triglyceride (TG) (47%), and LDL cholesterol (95%), together with lipid accumulation in the liver. Compared with the EtOH group, the EtOH + Lut group had significant reductions in serum concentrations of ALT (43%), TG (22%), LDL cholesterol (52%), and lipid accumulation in the liver. Ethanol elevated liver expression of lipogenic genes including sterol regulatory element-binding protein 1c (Srebp1c) (560%), fatty acid synthase (Fasn) (190%), acetyl-CoA carboxylase (Acc) (48%), and stearoyl-CoA desaturase 1 (Scd1) (286%). Luteolin reduced ethanol-induced expression of these genes in the liver: Srebp1c (79%), Fasn (80%), Acc (60%), and Scd1 (89%). In cultured hepatocytes, luteolin prevented alcohol-induced lipid accumulation and increase in the expression of lipogenic genes. The transcriptional activity of the master regulator of lipid synthesis, sterol regulatory element-binding protein (SREBP), was enhanced by ethanol treatment (160%) and reduced by luteolin administration (67%). In addition, ethanol-induced reduction of AMP-activated protein kinase and SREBP-1c phosphorylation was abrogated by luteolin. Collectively, our study indicates that luteolin is effective in ameliorating ethanol-induced hepatic steatosis and injury.
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Luteolin, analytical standard
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Luteolin, primary reference standard
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology