Merck
  • Potent inhibition of alcohol self-administration in alcohol-preferring rats by a κ-opioid receptor antagonist.
Potent inhibition of alcohol self-administration in alcohol-preferring rats by a κ-opioid receptor antagonist.
The Journal of pharmacology and experimental therapeutics (2014-05-13)
John R Cashman, Marc R Azar
ABSTRACT
A substituted aryl amide derivative of 6-naltrexamine--17-cyclopropylmethyl-3,14β-dihydroxy-4,5α-epoxy-6β-[(4'-trimethylfluoro)benzamido]morphinan-hydrochloride--(compound 5), previously shown to be a potent κ-opioid receptor antagonist, was used to characterize the physicochemical properties and efficacy to decrease alcohol self-administration in alcohol-preferring rats (P-rats) and binge-like P-rats. Previous studies showed that compounds closely related to compound 5 possessed favorable properties regarding penetration of the blood-brain barrier. Pharmacokinetic studies showed that compound 5 had acceptable bioavailability. In contrast to other κ-receptor antagonists, in particular norbinaltorphimine, compound 5 showed favorable drug-like properties. Based on these findings, further studies were done. Safety studies showed that compound 5 was not hepatotoxic at doses 200-fold greater than an efficacious dose. The effects of compound 5 or naltrexone on the hepatotoxicity of thiobenzamide were investigated. In contrast to naltrexone, which exacerbated thiobenzamide-mediated hepatotoxicity, compound 5 was observed to be hepatoprotective. Based on the physicochemical properties of compound 5, the compound was examined in rat animal models of alcohol self-administration. The inhibition of ethanol self-administration by compound 5 in alcohol-dependent and alcohol-nondependent P-rats trained to self-administer a 10% (w/v) ethanol solution, using operant techniques, showed very potent efficacy (i.e., estimated ED50 values of 4-5 μg/kg). In a binge-like P-rat animal model, inhibition of alcohol self-administration by compound 5 had an estimated ED50 value of 8 μg/kg. The results suggest that compound 5 is a potent drug-like κ-opioid receptor antagonist of utility in alcohol cessation medications development.
Product Description
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol, standard for GC
Supelco
Naltrexone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®