Merck
  • Right portal vein embolization with absolute ethanol in major hepatic resection for hepatobiliary malignancy.
Right portal vein embolization with absolute ethanol in major hepatic resection for hepatobiliary malignancy.
The British journal of surgery (2014-06-13)
K Sofue, Y Arai, K Shimada, Y Takeuchi, T Kobayashi, M Satake, K Sugimura
ABSTRACT
This study aimed to evaluate the safety and efficacy of preoperative right portal vein embolization (PVE) with absolute ethanol in patients with hepatobiliary malignancies. PVE was performed via a percutaneous transhepatic ipsilateral approach, and the right portal branch was embolized with absolute ethanol. Technical success and complications following PVE, and changes in liver enzyme levels were evaluated. Changes in future liver remnant (FLR) and FLR/total functional liver volume ratio were calculated. Complications following hepatic resection were assessed. A total of 83 patients with hepatobiliary malignancies (53 men, 30 women; mean age 68 years) underwent right PVE. Tumour types were hilar cholangiocarcinoma (37), liver metastases (14), gallbladder cancer (13), intrahepatic cholangiocellular carcinoma (10) and hepatocellular carcinoma (HCC) (9). PVE was performed successfully in all patients. Four patients (5 per cent) developed complications following PVE (liver abscess 2, left portal vein thrombosis 1, pseudoaneurysm 1), but this did not preclude hepatic resection. Liver enzyme levels rose transiently after PVE. The mean FLR and FLR/total functional liver volume increased after PVE (from 366 to 513 cm(3) and from 31 to 43 per cent respectively; both P < 0·001). Changes in the FLR and FLR/total functional liver volume ratio were comparable between patients with HCC and those with other malignancies (42 and 44 per cent, and 12 and 12 per cent, respectively). Sixty-nine of 83 patients underwent hepatic resection at a median of 25 days after PVE, with no postoperative mortality. Preoperative right PVE with absolute ethanol is safe and effective for induction of selective hepatic hypertrophy in patients with hepatobiliary malignancy.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material