Merck
  • Ethanol affects hepatitis C pathogenesis: humanized SCID Alb-uPA mouse model.
Ethanol affects hepatitis C pathogenesis: humanized SCID Alb-uPA mouse model.
Biochemical and biophysical research communications (2014-06-24)
Natalia A Osna, Kusum K Kharbanda, Yimin Sun, Ronda L Simpson, Larisa E Poluektova, Murali Ganesan, James L Wisecarver, David F Mercer
ABSTRACT
Alcohol consumption exacerbates the course of hepatitis C viral (HCV) infection, worsens outcomes and contributes to the development of chronic infection that exhibits low anti-viral treatment efficiency. The lack of suitable in vivo models makes HCV-ethanol studies very difficult. Here, we examine whether chimeric SCID Alb-uPA mice transplanted with human hepatocytes and infected with HCV develop worsening pathology when fed ethanol. After 5 weeks of feeding, such mice fed chow+water (control) or chow+20% ethanol in water (EtOH) diets mice developed oxidative stress, decreased proteasome activity and increased steatosis. Importantly, HCV(+) mice in the control group cleared HCV RNA after 5 weeks, while the infection persisted in EtOH-fed mice at the same or even higher levels compared with pre-feeding HCV RNA. We conclude that in chimeric SCID Alb-uPA mice, EtOH exposure causes the complex biochemical and histological changes typical for alcoholic liver injury. In addition, ethanol feeding delays the clearance of HCV RNA thereby generating persistent infection and promoting liver injury. Overall, this model is appropriate for conducting HCV-ethanol studies.
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard