Merck
  • Systematic review with meta-analysis: alcohol consumption and the risk of colorectal adenoma.
Systematic review with meta-analysis: alcohol consumption and the risk of colorectal adenoma.
Alimentary pharmacology & therapeutics (2014-06-20)
J-Z Zhu, Y-M Wang, Q-Y Zhou, K-F Zhu, C-H Yu, Y-M Li
ABSTRACT
Studies on the relation between alcohol consumption and risk of colorectal adenoma (CRA), a precursor of colorectal cancer, have been inconsistent. A systematic review with meta-analysis was conducted to investigate the association and the dose-response of alcohol with CRA. A literature search was performed on PubMed to identify relevant studies published up to January 2014. A fixed or random effects model was used to estimate summarised relative risks (RRs) and 95% confidence intervals (CIs) for the association between alcohol intake and CRA risk. Statistical heterogeneity between studies was assessed with the χ(2) statistic and quantified by I². Twenty-three case-control studies and two cohort studies were included in the meta-analysis. All drinkers were associated with 17% increased risk for CRA, compared with nondrinkers or occasional alcohol drinkers. The dose-response analysis demonstrated that for drinkers of 10, 25, 50 and 100 g/day alcohol consumption, the estimated RRs of CRA were 1.02 (95% CI 0.89-1.16), 1.06 (95% CI 0.92-1.20), 1.16 (95% CI 1.02-1.33) and 1.61 (95% CI 1.42-1.84) respectively, in comparison with non-/occasional drinkers. The risks were consistent in the subgroup analyses of gender and site of adenoma, while it was stronger in European studies than the studies in the US and Asia. This study suggests that alcohol intake is related to a significant increase of risk for colrectal adenoma.
Product Description
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol, standard for GC
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard