Merck
  • Early life stage (ELS) toxicity of sucralose to fathead minnows, Pimephales promelas.
Early life stage (ELS) toxicity of sucralose to fathead minnows, Pimephales promelas.
Bulletin of environmental contamination and toxicology (2014-08-15)
K I Stoddard, D B Huggett
ABSTRACT
Sucralose, an intense artificial sweetener, has been detected in wastewater and surface waters at concentrations ranging from ng/L to low µg/L. Although over a hundred studies have been conducted to evaluate the safety of sucralose for human consumption, few studies have focused on the chronic ecotoxicological effects of this compound in fish. As a remedy to this data gap, an early-life stage toxicity test was conducted to assess the effects of sucralose on hatching, survival, and growth of fathead minnows (Pimephales promelas). Hatching, survival, and growth were unaffected by 98 mg/L of sucralose. The Lowest-Observed-Effect Concentration (LOEC) and the No-Observed-Effect Concentration (NOEC) for fathead minnows determined by this study are >98 and 98 mg/L, respectively. The results from this study suggest that the concentrations of sucralose detected in the environment are well below those required to cause adverse effects to developing aquatic organisms.
Sigma-Aldrich
Ethyl 3-aminobenzoate methanesulfonate, 98%
Supelco
Ethyl 3-aminobenzoate methanesulfonate salt, analytical standard
Supelco
Sucralose, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sucralose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucralose, analytical standard
USP
Sucralose, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucralose, ≥98.0% (HPLC)
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Supelco
Acetonitrile, analytical standard
Sucrose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%