Merck
  • Thrombin inhibition with dabigatran protects against high-fat diet-induced fatty liver disease in mice.
Thrombin inhibition with dabigatran protects against high-fat diet-induced fatty liver disease in mice.
The Journal of pharmacology and experimental therapeutics (2014-08-21)
Anna K Kopec, Nikita Joshi, Keara L Towery, Karen M Kassel, Bradley P Sullivan, Matthew J Flick, James P Luyendyk
ABSTRACT
Nonalcoholic fatty liver disease (NAFLD) is the hepatic manifestation of obesity and metabolic syndrome. Robust coagulation cascade activation is common in obese patients with NAFLD. We identified a critical temporal relationship between thrombin generation and the manifestation of hepatic steatosis, inflammation, and injury in C57BL/6J mice fed a high-fat diet (HFD) for 1, 2, and 3 months. Mice fed a HFD exhibited dramatic increases in hepatocellular injury and inflammation over time. Hepatic fibrin deposition preceded an increase in serum alanine aminotransferase, and the most dramatic changes in liver histopathology occurred in conjunction with a detectable increase in plasma thrombin-antithrombin levels at 3 months. To directly determine whether thrombin activity promotes NAFLD pathogenesis, mice were fed a HFD and simultaneously treated with the direct thrombin inhibitor dabigatran etexilate for 3 months. Notably, dabigatran treatment significantly reduced hepatic fibrin deposition, hepatic inflammation, hepatocellular injury, and steatosis in mice fed a HFD. Of interest, dabigatran treatment also significantly attenuated HFD-induced body weight gain. Gene expression analysis suggested that thrombin potentially drives NAFLD pathogenesis by altering the expression of genes associated with lipid metabolism and bile acid synthesis. Collectively, the results suggest that thrombin activity is central to HFD-induced body weight gain, liver injury, and inflammation and provide the proof-of-principle evidence that pharmacological thrombin inhibition could be effective in limiting NAFLD and associated pathologies.
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Fibrin from human plasma, insoluble powder
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Supelco
Beta Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-Alanine, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
β-Alanine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
2-Methylbutane, anhydrous, ≥99%
Sigma-Aldrich
β-Alanine, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
β-Alanine, 99%
Supelco
2-Methylbutane, analytical standard
Sigma-Aldrich
2-Methylbutane, suitable for HPLC, ≥99.5%
Supelco
β-Alanine, analytical standard
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%