Merck
  • Urban overdose hotspots: a 12-month prospective study in Dublin ambulance services.
Urban overdose hotspots: a 12-month prospective study in Dublin ambulance services.
The American journal of emergency medicine (2014-08-27)
Jan Klimas, Martin O'Reilly, Mairead Egan, Helen Tobin, Gerard Bury
ABSTRACT
Opioid overdose (OD) is the primary cause of death among drug users globally. Personal and social determinants of overdose have been studied before, but the environmental factors lacked research attention. Area deprivation or presence of addiction clinics may contribute to overdose. The objective of the study is to examine the baseline incidence of all new ODs in an ambulance service and their relationship with urban deprivation and presence of addiction services. A prospective chart review of prehospital advanced life support patients was performed on confirmed OD calls. Demographic, geographic, and clinical information, that is, presentation, treatment, and outcomes, was collected for each call. The census data were used to calculate deprivation. Geographical information software mapped the urban deprivation and addiction services against the overdose locations. There were 469 overdoses, 13 of which were fatal; most were male (80%), of a young age (32 years), with a high rate of repeated overdoses (26%) and common polydrug use (9.6%). Most occurred in daytime (275) and on the streets (212). Overdoses were more likely in more affluent areas (r = .15; P < .05) and in a 1000-m radius of addiction services. Residential overdoses were in more deprived areas than street overdoses (mean difference, 7.8; t170 = 3.99; P < .001). Street overdoses were more common in the city center than suburbs (χ(2)(1) = 33.04; P < .001). The identified clusters of increased incidence-urban overdose hotspots-suggest a link between environment characteristics and overdoses. This highlights a need to establish overdose education and naloxone distribution in the overdose hotspots.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, absolute, sales not in Germany, ≥99.8% (vol.)
Sigma-Aldrich
Ethanol
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®