Merck
  • Interarytenoid sodium carboxymethylcellulose gel injection for management of pediatric aspiration.
Interarytenoid sodium carboxymethylcellulose gel injection for management of pediatric aspiration.
The Annals of otology, rhinology, and laryngology (2014-06-26)
David L Horn, Kim DeMarre, Sanjay R Parikh
ABSTRACT
This study aimed to investigate the role of interarytenoid injection laryngoplasty (IL) for the management of pediatric aspiration. Medical records of 30 patients, 9 female, with radiographically confirmed chronic aspiration who underwent intraoperative IL were retrospectively reviewed. Clinical improvement was defined as successful advancement of feeds to thinner consistencies. Clinical improvement was observed in 57% of patients. Six children with type 1 posterior laryngeal cleft (PLC-1) were not significantly more likely to show improvement compared to the children without PLC-1. Type 1 posterior laryngeal cleft was associated with older age and higher prevalence of neurodevelopmental risk factors relative to absence of PLC-1. Patients with PLC-1 were more likely than noncleft patients to show recurrence of symptoms after initial improvement with IL. Five patients underwent endoscopic repair. Repair was successful in 3 patients who improved after IL but not in 2 patients who did not improve after IL. Chronic aspiration can improve after IL even in patients with normal anatomy. Injection laryngoplasty can be performed to improve selection of PLC-1 patients for definitive endoscopic repair. Further prospective research, with a randomized control group, is needed to understand whether interarytenoid incompetence plays a role in some patients with chronic aspiration, who do not have a PLC-1.
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.9
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 2500-6000 cP 
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 50-200 cP , c=4% H2O at 25­°C
Sigma-Aldrich
Carboxymethylcellulose sodium, meets USP testing specifications, Medium viscosity
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.7
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 1.2
Sigma-Aldrich
Carboxymethylcellulose sodium salt, medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, low viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, ultra high viscosity, highly purified
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity