Merck
  • STIM1 phosphorylation triggered by epidermal growth factor mediates cell migration.
STIM1 phosphorylation triggered by epidermal growth factor mediates cell migration.
Biochimica et biophysica acta (2014-12-03)
Vanessa Casas-Rua, Patricia Tomas-Martin, Aida M Lopez-Guerrero, Ignacio S Alvarez, Eulalia Pozo-Guisado, Francisco Javier Martin-Romero
ABSTRACT
STIM1 is a key regulator of store-operated calcium entry (SOCE), and therefore a mediator of Ca²⁺ entry-dependent cellular events. Phosphorylation of STIM1 at ERK1/2 target sites has been described as enhancing STIM1 activation during intracellular Ca²⁺ emptying triggered by the inhibition of the sarco(endo)plasmic Ca²⁺ -ATPase with thapsigargin. However, no physiological function is known for this specific phosphorylation. The present study examined the role of STIM1 phosphorylation in cell signaling triggered by EGF. Using a human endometrial adenocarcinoma cell line (Ishikawa cells) EGF or H-Ras(G12V), an active mutant of H-Ras, was found to trigger STIM1 phosphorylation at residues Ser575, Ser608, and Ser621, and this process was sensitive to PD0325901, an inhibitor of ERK1/2. Both, ERK1/2 activation and STIM1 phosphorylation took place in the absence of extracellular Ca²⁺, indicating that both events are upstream steps for Ca²⁺entry activation. Also, EGF triggered the dissociation of STIM1 from EB1 (a regulator of microtubule plus-ends) in a manner similar to that reported for the activation of STIM1 by thapsigargin. Migration of the Ishikawa cells was impaired when STIM1 phosphorylation was targeted by Ser-to-Ala substitution mutation of ERK1/2 target sites. This effect was also observed with the Ca²⁺ channel blocker SKF96365. Phosphomimetic mutation of STIM1 restored the migration to levels similar to that found for STIM1-wild type. Finally, the increased vimentin expression and relocalization of E-cadherin triggered by EGF were largely inhibited by targeting STIM1 phosphorylation, while STIM1-S575E/S608E/S621E normalized the profiles of these two EMT markers.
Product Description
SAFC
BIS-TRIS
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Diphenylamine, ReagentPlus®, 99%
Sigma-Aldrich
Diphenylamine, ACS reagent, ≥99%
Sigma-Aldrich
Diphenylamine, puriss. p.a., suitable for redox indicator, ACS reagent, reag. Ph. Eur., ≥98% (GC)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Supelco
Diphenylamine, PESTANAL®, analytical standard
SAFC
BIS-TRIS
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Supelco
Diphenylamine solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Epidermal Growth Factor from murine submaxillary gland, EGF, suitable for cell culture
Sigma-Aldrich
SKF-96365, ≥98% (HPLC), solid